Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Scoops

25 ml, 70 mm, 30 mm
Catalog # 231-0359
Supplier:   NICKEL ELECTRO
Scoops
Catalog # 231-0359
Supplier:  NICKEL ELECTRO
Scoops
Scoops

25 ml, 70 mm, 30 mm

Price
SFr. 60.00
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel. Round bottom. Ideal for use as weighing boats etc.

  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Specifications
  • Width:
    30 mm
  • Capacity:
    25 ml
  • Length:
    70 mm