Staining dishes, WHEATON®
Staining dish, complete, includes dish, cover and rack, 95×76×64 mm
Catalog # 631-2500
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Soda-lime glass and stainless steel.
- Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
WHEA900200_TDS_EN
Datasheet
Specifications
- Description:Staining dish, complete, includes dish, cover and rack
- W×D×H:95×76×64 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



