Syringes
Syringe, glass, metal centre nozzle, Luer slip, grad. 4,0 ml, 50 ml
613-4515
: MURRAY S
Glass syringes with metal centre nozzle, without needles.
- Centre nozzle with Luer slip fitting
- Volume:50 ml
- Description:Syringe, glass, metal centre nozzle, Luer slip, grad. 4,0 ml



