Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Cryogenic vials, Cryovial®

5,00 ml, 12,5×91 mm, Yes
Catalog # 720-1002
Supplier:   Simport Scientific
Cryogenic vials, Cryovial®
Catalog # 720-1002
Supplier:  Simport Scientific
Cryogenic vials, Cryovial®
Cryogenic vials, Cryovial®

5,00 ml, 12,5×91 mm, Yes

Price
SFr. 851.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

PP vial and closure. Designed for the storage of biological material, human or animal cells, at temperatures as low as −196 °C (but should be used only in the gas phase of liquid nitrogen).

  • Certified RNase-, DNase-, pyrogen- and DNA-free
  • With graduations and marking area

Round bottom vials can be centrifuged up to 17000×g

Round bottom vials can be centrifuged up to 17 000 ×g

Specifications
  • Cap included:
    Yes
  • Ø×H:
    12,5×91 mm
  • Capacity:
    5,00 ml
  • Seq:
    16