HTS 24-multiwell insert system, Falcon®
The Falcon® HTS 24-multiwell insert system is designed to automate many commonly used membrane-based cell assays and increase the productivity and throughput of these assays in the drug discovery process. Available in a choice of membrane pore sizes (1, 3 and 8 µm), 24-multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability and absorption studies (Caco-2 cells), chemotaxis, cell migration and invasion assays.
- All the benefits of individual cell culture inserts in an automation-friendly format that is compatible with most robotic and fluid handling equipment
- Handle 24 inserts simultaneously, as all 24 inserts are part of a single unit that is compatible with Falcon® 24-well plates and feeder tray
Pore density: 1 µm insert (pores/cm²): 1,6×10⁶
Pore density: 3 µm insert (pores/cm²): 8,0×10⁵
Pore density: 8 µm insert (pores/cm²): 1,0×10⁵
- Pore size:3 µm
- Description:With 24-well plate and lid
- Cat. No.:734-0380
- Effective growth area of membrane:0,3 cm²
- Suggested media volume in well:1000 - 4000 µl
- Insert height:18 mm
- Suggested media volume in insert:300 - 500 µl
- Effective diameter of membrane:6,5 mm
- Distance from membrane to bottom of well:2,0 mm
- Effective growth area in 24-well plate:2,0 cm²
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



