TACROL053 Printer paper roll 1 * 5 items
Catalog # PRIOZZZ/ROL/005
Anbieter: Priorclave
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Vacuum seal PTFE / FKM
Anbieter: Heidolph Instruments GmbH & Co.KG
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Petri dishes, Star™ Dish
Anbieter: PHOENIX BIOMEDICAL
VWR®, Accessories for Mechanical Pipettes
Anbieter: VWR Collection
Accessories for PIPETBOY acu / pro
Anbieter: Integra
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Fine dosage syringes, Omnifix®-F Solo
Anbieter: B. Braun
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
VWR®, Nitrile Gloves
Anbieter: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Anbieter: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Anbieter: VWR Collection
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Anbieter: EPPENDORF
epT.I.P.S.® Reloads, Pipette Tips
Anbieter: EPPENDORF