Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Burette, Sofnol, auto zero, MBL®

Burette, Sofnol, auto zero, MBL®

10 ml, 0,1 ml, 0,10 ± ml
Catalog # BIBBBRB004
Supplier:   SciLabware
Burette, Sofnol, auto zero, MBL®
Catalog # BIBBBRB004
Supplier:  SciLabware
Burette, Sofnol, auto zero, MBL®
Burette, Sofnol, auto zero, MBL®

10 ml, 0,1 ml, 0,10 ± ml

Price
SFr. 155.00
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass, class B.

  • Blue enamel graduations
  • Easy filling and automatic zero
  • With simple rubber tube and pinch valve for low cost replacement

The burette can be filled by squeezing the bottle.

Complies with ISO 385

Specifications
  • Capacity:
    10 ml
  • Seq:
    01
  • Division:
    0,1 ml
  • Tolerance:
    0,10 ± ml
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Rubber pinch clip and glass jet
    Rubber pinch clip and glass jet
    BIBBBRB042
    Log in for availability
    1 * 5 ST - SFr. 88.90