Orders LinkContactUsLinkComponent
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
55 mm, 3, 24/29 NS
Catalog # 532-0097
Supplier:   SciLabware
Büchner funnels, Quickfit®
Catalog # 532-0097
Supplier:  SciLabware
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
55 mm, 3, 24/29 NS
Price
SFr. 141.00
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.
Specifications
  • Cone size:
    24/29 NS
  • Plate Ø:
    55 mm
  • Porosity:
    3