Orders LinkContactUsLinkComponent
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
-, S 35
Catalog # BIBBFS35UB
Supplier:   SciLabware
Spherical ground joints, Quickfit®
Catalog # BIBBFS35UB
Supplier:  SciLabware
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
-, S 35
Price
SFr. 38.80
Log in for availability
1 * 1 ST
Product Details & Documents
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Specifications
  • Ball:
    -
  • Socket:
    S 35