Hardness (Calcium and Total, Sequential) Reagent Sets, for Digital Titrator, ~ 100 tests, MR 10-4000 mg/L as CaCO3, without Sulfuric Acid 0.0800 M and 0.800 M titrants) 1 * 1 items
Catalog # HACH2272100
Supplier: Hach
Frequently Bought Together