Orders LinkContactUsLinkComponent
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, Thoma
Catalog # 631-0697
Supplier:   MARIENFELD
Counting chambers, Thoma
Catalog # 631-0697
Supplier:  MARIENFELD
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, Thoma
Price
SFr. 62.80
Log in for availability
1 * 1 ST
Product Details & Documents
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.
Specifications
  • Description:
    Counting chamber, dark lines, Thoma
  • Seq:
    2