Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Trypsin | Avantor

Trypsin

Trypsin 0,25% in PBS, without calcium, without magnesium, without phenol red, 100 ml
Catalog # L0910-100
Supplier:   Biowest
Trypsin
Catalog # L0910-100
Supplier:  Biowest
CAS Number:  
Trypsin
Trypsin

Trypsin 0,25% in PBS, without calcium, without magnesium, without phenol red, 100 ml

CAS Number:  
Price
SFr. 14.90
Log in for availability
1 * 100 mL
Product Details & Documents

Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Trypsin 0,25% in PBS, without calcium, without magnesium, without phenol red

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..