Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Volumetric flasks, class A

Volumetric flasks, class A

1000 ml, 24/29, 310 mm, 0,40 ± ml
Catalog # HIRS7570193
Supplier:   HIRSCHMANN
Volumetric flasks, class A
Catalog # HIRS7570193
Supplier:  HIRSCHMANN
Volumetric flasks, class A
Volumetric flasks, class A

1000 ml, 24/29, 310 mm, 0,40 ± ml

Price
SFr. 45.60
Log in for availability
1 * 1 ST
Product Details & Documents

PMP, translucent, with PP NS-stopper.

Specifications
  • NS:
    24/29
  • Capacity:
    1000 ml
  • Height:
    310 mm
  • Tolerance:
    0,40 ± ml