Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Volumetric flasks, class A

Volumetric flasks, class A

25 ml, 10/19, 115 mm, 0,04 ± ml
Catalog # HIRS7570170
Supplier:   HIRSCHMANN
Volumetric flasks, class A
Catalog # HIRS7570170
Supplier:  HIRSCHMANN
Volumetric flasks, class A
Volumetric flasks, class A

25 ml, 10/19, 115 mm, 0,04 ± ml

Price
SFr. 41.80
Log in for availability
1 * 2 ST
Product Details & Documents

PMP, translucent, with PP NS-stopper.

Specifications
  • NS:
    10/19
  • Capacity:
    25 ml
  • Height:
    115 mm
  • Tolerance:
    0,04 ± ml