Solvent filter/degasser
Funnel base, glass, PTFE
Catalog # HEWL5188-2746
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
- Degasses eluents as particulates are removed
- Increases solvent inlet lifetime
- Eliminates pump downtime caused by air locks and particulates in check valves
- Decreases piston wear, while increasing column life
Specifications
- Description:Funnel base, glass
- Material:PTFE
- Seq:01