Methyl decanoate
Glass bottle for solids, 500 mg
Catalog # BTRN1261
Jump to:
Discontinued
Product Details & Documents
Synonyms: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
- CAS Number:110-42-9
- Formula:C₁₁H₂₂O₂
- MDL Number:MFCD00009580
- Molecular Weight:186.29 g/mol
- Boiling Pt:224 °C (1013 hPa)
- Melting Pt:–18 °C
- Density:0,8724 g/cm³ (20 °C)
- Flash Pt:94 °C
- EINECS:203-766-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



