Methyl enantate
Glass bottle for solids, 1 g
Catalog # BTRN1197
Jump to:
Discontinued
Product Details & Documents
Synonyms: Methyl Heptanoate, Methyl Enanthate, Enanthic acid methyl ester, Heptanoic acid methyll ester, Heptylic acid methyl ester, Heptanoic acid methyl ester, Enanthic acid methyll ester, Methyl heptanoate
- CAS Number:106-73-0
- Formula:C₈H₁₆O₂
- MDL Number:MFCD00009537
- Molecular Weight:144.21 g/mol
- Merck Index:12,04695
- Boiling Pt:172 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:0.88 g/cm³ (20 °C)
- EINECS:203-428-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



