Methyl tridecanoate
Glass bottle for solids, 100 mg
Catalog # BTRN1162
Jump to:
Discontinued
Product Details & Documents
Synonyms: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- CAS Number:1731-88-0
- Formula:C₁₄H₂₈O₂
- MDL Number:MFCD00008977
- Molecular Weight:228.38 g/mol
- Boiling Pt:290 °C (1013 hPa)
- Melting Pt:5.5 °C
- Density:0.864 g/cm³ (25 °C)
- EINECS:217-054-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



