5α-Cholestane
Glass bottle for solids, 100 mg
BTRN1115
- :481-21-0
- :C₂₇H₄₈
- :MFCD00066412
- :372.68 g/mol
- :12,02254
- :441 °C (1013 hPa)
- :79 °C
- :0.9133 g/cm³ (80 °C)
- :Density: 0.9133 g/cm³ (80 °C)
- :207-562-8
- Pack type:Glass bottle for solids
- Pk:100 mg