Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl arachidate

Methyl arachidate

Glass bottle for solids, 500 mg
Catalog # BTRN1031
Methyl arachidate
Catalog # BTRN1031
Supplier:  
CAS Number:  1120-28-1
Methyl arachidate

Glass bottle for solids, 500 mg

CAS Number:  1120-28-1
Discontinued
Product Details & Documents
Synonyms:  Methyl eicosanoate, Methyl Arachidate
  • CAS Number:1120-28-1
  • Formula:C₂₁H₄₂O₂
  • MDL Number:MFCD00009014
  • Molecular Weight:326.56 g/mol
  • EINECS:214-304-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications