Nonadecanoic acid methyl ester
Glass bottle for solids, 100 mg
Catalog # BTRN1029
Jump to:
Discontinued
Product Details & Documents
Synonyms: Nonadecanoic acid methyll ester, Methyl Nonadecanoate
- CAS Number:1731-94-8
- Formula:C₂₀H₄₀O₂
- MDL Number:MFCD00009011
- Molecular Weight:312.54 g/mol
- EINECS:217-056-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



