Oleic acid
Glass bottle for solids, 1 g
BTRN1022
: (9Z)-9-Octadecenoic acid
- :112-80-1
- :C₁₈H₃₄O₂
- :MFCD00064242
- :282.47 g/mol
- :13,06898
- :286 °C (1013 hPa)
- :16.7 °C
- :0.8946 g/cm³ (20 °C)
- :180 °C
- :Freezer
- :204-007-1
- Pack type:Glass bottle for solids
- Pk:1 g