ContactUsLinkComponent
2ml wide mouth screw top clear vial fitted with green cap-rubber/ptfe line
Catalog # CRMA2-SVWCG8
Supplier:  CHROMACOL
2ml wide mouth screw top clear vial fitted with green cap-rubber/ptfe line
Catalog # CRMA2-SVWCG8
Supplier:  CHROMACOL