ContactUsLinkComponent
Flat display cases for 20 microscope slides. Constructed from strong cardboard with individual cut outs and cover. Model PM20V has an additional fastening. 1 * 1 items
Catalog # LIEDPM20V
Supplier:  LIEDER JOHANNES
Flat display cases for 20 microscope slides. Constructed from strong cardboard with individual cut outs and cover. Model PM20V has an additional fastening. 1 * 1 items
Catalog # LIEDPM20V
Supplier:  LIEDER JOHANNES
Frequently Bought Together