ContactUsLinkComponent
I 800 DIN,COMBINED IODIDE SELECTIVE ELECTRODE, WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0.006 TO 127,000 MG/L, REQUIRES ELY/BR/503 REFEREN CE ELECTROLYTE, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I DELIVERY CONTAINS ELECTROLYTE 1 * 1 items
Catalog # 662-7381
Supplier:  WTW
I 800 DIN,COMBINED IODIDE SELECTIVE ELECTRODE, WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0.006 TO 127,000 MG/L, REQUIRES ELY/BR/503 REFEREN CE ELECTROLYTE, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I DELIVERY CONTAINS ELECTROLYTE 1 * 1 items
Catalog # 662-7381
Supplier:  WTW
Frequently Bought Together