ContactUsLinkComponent
I 500 BNC,IODIDE SELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, BNC PLUG, 0.006 TO 127,000 MG/L, REQUIRES REFERENCE R 503/D OR /P WITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I. ACCESSORIES ARE NOT INCLUDED IN DELIVERY 1 * 1 items
Catalog # WTWA106618
Supplier:  WTW
I 500 BNC,IODIDE SELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, BNC PLUG, 0.006 TO 127,000 MG/L, REQUIRES REFERENCE R 503/D OR /P WITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I. ACCESSORIES ARE NOT INCLUDED IN DELIVERY 1 * 1 items
Catalog # WTWA106618
Supplier:  WTW
Frequently Bought Together