Orders LinkContactUsLinkComponent
Spherical ground joints, Quickfit® | Avantor
Spherical ground joints, Quickfit®
-, S 19
Katalognr. 201-8207
Anbieter:   SciLabware
Spherical ground joints, Quickfit®
Katalognr. 201-8207
Anbieter:  SciLabware
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
-, S 19
Preis
SFr. 256.00
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Technische Daten
  • Ball:
    -
  • Socket:
    S 19