Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl octanoate

Methyl octanoate

Glass bottle for solids, 1 g
Catalog # MAYA1199
Supplier:   MATREYA
Methyl octanoate
Catalog # MAYA1199
Supplier:  MATREYA
CAS Number:  111-11-5
Methyl octanoate

Glass bottle for solids, 1 g

CAS Number:  111-11-5
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
  • CAS Number:111-11-5
  • Formula:C₉H₁₈O₂
  • MDL Number:MFCD00009551
  • Molecular Weight:158.24 g/mol
  • Boiling Pt:190 °C (1013 hPa)
  • Melting Pt:–36,7 °C
  • Melting Pt:–36.7 °C
  • Density:0,877 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • EINECS:203-835-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.