Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Methyl tridecanoate

Glass bottle for solids, 100 mg
Catalog # MAYA1162
Supplier:   MATREYA
Methyl tridecanoate
Catalog # MAYA1162
Supplier:  MATREYA
CAS Number:  1731-88-0
Methyl tridecanoate

Glass bottle for solids, 100 mg

CAS Number:  1731-88-0
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Tridecylic acid methyl ester, Tridecanoic acid methyl ester
  • CAS Number:1731-88-0
  • Formula:C₁₄H₂₈O₂
  • MDL Number:MFCD00008977
  • Molecular Weight:228.38 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:5.5 °C
  • Density:0.864 g/cm³ (25 °C)
  • EINECS:217-054-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications