Methyl arachidate
Glass bottle for solids, 500 mg
Catalog # MAYA1031
Supplier: MATREYA
Jump to:
Product Details & Documents
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- EINECS:214-304-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



