Stearic acid
Glass bottle for solids, 1 g
Catalog # MAYA1020
Supplier: MATREYA
Jump to:
Product Details & Documents
Synonyms: Octadecanoic acid, Octadecylic acid
- CAS Number:57-11-4
- Formula:C₁₈H₃₆O₂
- MDL Number:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Boiling Pt:361 °C (1013 hPa)
- Melting Pt:67 °C
- Density:0.94 g/cm³ (20 °C)
- Flash Pt:196 °C (closed cup)
- Storage Temperature:Refrigerator
- EINECS:200-313-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



