ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat IL-1beta. Anti-Rat IL-1beta specific antibody wa s purified by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P80BT-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat IL-1beta. Anti-Rat IL-1beta specific antibody wa s purified by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P80BT-1
Supplier:  PeproTech, Inc.
Frequently Bought Together