ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-4. Anti-Murine IL-4 specific antibody was purifi ed by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P54BT-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-4. Anti-Murine IL-4 specific antibody was purifi ed by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P54BT-1
Supplier:  PeproTech, Inc.
Frequently Bought Together