ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-2. Anti-Human IL-2 specific antibody was purifie d by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P22BT-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-2. Anti-Human IL-2 specific antibody was purifie d by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P22BT-1
Supplier:  PeproTech, Inc.
Frequently Bought Together