ContactUsLinkComponent
Produced from sera of goats pre-immunized with highly pure (>98%) recombinant murine SDF-1alpha. Anti-Murine SDF-1alpha specific ant ibody was purified by affinity chromatography employing immobilized mSDF-1alpha matrix. 1 * 1 mg
Catalog # PEPR500-P164G-1
Supplier:  PeproTech, Inc.
Produced from sera of goats pre-immunized with highly pure (>98%) recombinant murine SDF-1alpha. Anti-Murine SDF-1alpha specific ant ibody was purified by affinity chromatography employing immobilized mSDF-1alpha matrix. 1 * 1 mg
Catalog # PEPR500-P164G-1
Supplier:  PeproTech, Inc.
Frequently Bought Together