ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hMIP-3. Anti-Human MIP-3 specific antibody was purif ied by affinity chromatography employing immobilized hMIP-3 matrix. 1 * 1 mg
Catalog # PEPR500-P124-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hMIP-3. Anti-Human MIP-3 specific antibody was purif ied by affinity chromatography employing immobilized hMIP-3 matrix. 1 * 1 mg
Catalog # PEPR500-P124-1
Supplier:  PeproTech, Inc.
Frequently Bought Together