ContactUsLinkComponent
Synonyms: CXCL11 B-R1 Description: I-TAC is a 'non-ELR' CXC chemokine that is regulated by interferon and signals through the CXCR3 receptor. I-TAC is chemoattractant for IL-2 activated T cells, but does not affect freshly isolated un-stimulated T i 1 * 1 mg
Catalog # PEPR300-46-1
Supplier:  PeproTech, Inc.
Synonyms: CXCL11 B-R1 Description: I-TAC is a 'non-ELR' CXC chemokine that is regulated by interferon and signals through the CXCR3 receptor. I-TAC is chemoattractant for IL-2 activated T cells, but does not affect freshly isolated un-stimulated T i 1 * 1 mg
Catalog # PEPR300-46-1
Supplier:  PeproTech, Inc.
Frequently Bought Together