Orders LinkContactUsLinkComponent
Software for data loggers, elproLOG ANALYZE
Software for data loggers, elproLOG ANALYZE
elproLOG ANALYZE software, full version
Catalog # ELPR800397
Supplier:   ELPRO-BUCHS
Software for data loggers, elproLOG ANALYZE
Catalog # ELPR800397
Supplier:  ELPRO-BUCHS
Software for data loggers, elproLOG ANALYZE
elproLOG ANALYZE software, full version
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
elproLOG ANALYZE is a software for all elpro data loggers. It provides all functions such as data analysis, reporting and configuration of data loggers.
  • User friendly
  • Displays statistics on measurement data
  • Optional audit trail
  • Multiple zoom function
Specifications
  • Description:
    elproLOG ANALYZE software, full version
  • Seq:
    01