Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pestles

Pestles

55 mm, 210 mm, 410-9124, 410-9114
Catalog # 410-0079
Pestles
Catalog # 410-0079
Pestles
Pestles

55 mm, 210 mm, 410-9124, 410-9114

Price
SFr. 110.00
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

Certifications: Manufactured to DIN 12906.

Specifications
  • For:
    410-9124, 410-9114
  • Head Ø:
    55 mm
  • Length:
    210 mm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.