Pestles
48 mm, 185 mm, 410-9113
410-0078
Porcelain with glazed handle, glazed or unglazed grinding surface.
Certifications: Manufactured to DIN 12906.
- For:410-9113
- Head Ø:48 mm
- Length:185 mm
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



