Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Pestles

36 mm, 150 mm, 410-9110
Catalog # 410-0076
Pestles
Catalog # 410-0076
Pestles
Pestles

36 mm, 150 mm, 410-9110

Price
69.60 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

Specifications
  • For:
    410-9110
  • Head Ø:
    36 mm
  • Length:
    150 mm
  • Seq:
    04