Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Temperature indicating strips

Temperature indicating strips

116…154 °C
Catalog # TEST0646.1724
Supplier:   Testo
Temperature indicating strips
Catalog # TEST0646.1724
Supplier:  Testo
Temperature indicating strips
Temperature indicating strips

116…154 °C

Price
SFr. 38.00
Log in for availability
1 * 10 ST
Product Details & Documents

testoterm measuring strips are self-adhesive films with heat-sensitive elements for checking and monitoring temperature.

  • Change in colour in two to three seconds
  • Practical booklet format including 10 thermometer strips
  • 8 temperature levels

They are used for measurements on moving and/or small parts, for long-term monitoring.

Specifications
  • Seq:
    03
  • Range:
    116…154 °C
  • Cat. No.:
    TEST0646.1724
  • Storage temperature:
    +25 °C max.; Storage in refrigerator recommended