Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type

Sedimentation cones, Imhoff type

Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml, without stopcock
Catalog # LENZ05670003
Supplier:   Lenz Laborglas
Sedimentation cones, Imhoff type
Catalog # LENZ05670003
Supplier:  Lenz Laborglas
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type

Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml, without stopcock

Price
259.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Made of borosilicate glass 3.3.

  • White graduations
  • With or without stopcock
  • Excellent chemical and temperature resistance

Documents

Specifications
  • Type:
    without stopcock
  • Description:
    Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml
  • Cat. No.:
    LENZ05670003
  • Graduation:
    0 - 2 ml
  • Division:
    0,1 ml