Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 250-300 ml, 19/26
Catalog # 528-1203
Supplier:   Lenz Laborglas
Dissolved oxygen bottles
Catalog # 528-1203
Supplier:  Lenz Laborglas
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 250-300 ml, 19/26
Price
SFr. 55.60
Log in for availability
1 * 1 ST
Product Details & Documents
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Specifications
  • Capacity:
    250-300 ml
  • Description:
    Oxygen bottles acc. to Winkler
  • NS stopper:
    19/26
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 14/23 for Dissolved Oxygen Bottles
    Clamps NS 14/23 for Dissolved Oxygen Bottles
    705-0564
    Log in for availability
    1 * 1 ST - SFr. 11.70
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 19/26 for Dissolved Oxygen Bottles
    Clamps NS 19/26 for Dissolved Oxygen Bottles
    705-0565
    Log in for availability
    1 * 1 ST - SFr. 13.20
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..