Lovibond rybn colour 2.1r 11.0y 0.5n(5¼) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0051
Supplier: Lovibond Tintometer
Lovibond rybn colour 2.1r 11.0y 0.5n(5¼) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0051
Supplier: Lovibond Tintometer
Frequently Bought Together
Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Sucrose, Reference Standard
Supplier: USP
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
2,3-Dimethylbutane ≥98%
Supplier: Thermo Fisher Scientific