Lovibond RYBN 1.4R 7.3Y 0.2N (5 1/4) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0050
Supplier: Lovibond Tintometer
Lovibond RYBN 1.4R 7.3Y 0.2N (5 1/4) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0050
Supplier: Lovibond Tintometer
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Phenol red Reag. Ph. Eur.
Supplier: Merck
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
N-Nitroso-ethyl methylamine 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Pyraclofos certified 1 * 50 mg
Supplier: EHRENSTORFER
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
VWR®, Colour standard solutions ASTM
Supplier: VWR Chemicals
Benzocaine, Reference Standard
Supplier: USP
pH buffer solutions
Supplier: Hach
Supelco® TraceCERT® Organic Reference Standard, Propamocarb
Supplier: Merck
pH buffer solutions, colour coded
Supplier: Hach
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck