Lovibond rybn colour 0.8r 2.0y 0.1n(5¼)Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0048
Supplier: Lovibond Tintometer
Lovibond rybn colour 0.8r 2.0y 0.1n(5¼)Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0048
Supplier: Lovibond Tintometer
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ