Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Double spatulas, narrow

Double spatulas, narrow

Flexible, 250 mm, 11 mm
Catalog # RSGA030.250
Supplier:   RSG
Double spatulas, narrow
Catalog # RSGA030.250
Supplier:  RSG
Double spatulas, narrow
Double spatulas, narrow

Flexible, 250 mm, 11 mm

Price
SFr. 58.30
Log in for availability
1 * 5 ST
Product Details & Documents

Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.

Specifications
  • Description:
    Flexible
  • Length:
    250 mm
  • Blade width:
    11 mm