Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spoon spatulas, flat

Spoon spatulas, flat

Double spoon spatula, flat endings, 210 mm, 10 mm
Catalog # RSGA038.215
Supplier:   RSG
Spoon spatulas, flat
Catalog # RSGA038.215
Supplier:  RSG
Spoon spatulas, flat
Spoon spatulas, flat

Double spoon spatula, flat endings, 210 mm, 10 mm

Price
SFr. 36.50
Log in for availability
1 * 5 ST
Product Details & Documents

Flat spoon spatulas made of stainless steel. Available singe or double ended.

Specifications
  • Description:
    Double spoon spatula, flat endings
  • Length:
    210 mm
  • Blade width:
    10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.