Double spatulas, broad
400 mm, 30 mm
Catalog # RSGA092.400
Supplier: RSG
Jump to:
Product Details & Documents
Double spatulas with wide, flat endings. Made of stainless steel.
Documents
RSGA092.500_TDS_EN
Datasheet
Specifications
- Length:400 mm
- Blade width:30 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



